(S)-tert-Butyl 2-((tert-butoxycarbonyl)amino)-4-iodobutanoate 97%

Supplier: Ambeed
Warning

A386487-250MG A386487-1G
77251-976EA 156 USD
77251-976 77551-144
(S)-tert-Butyl 2-((tert-butoxycarbonyl)amino)-4-iodobutanoate 97%
(S)-tert-Butyl 2-((tert-butoxycarbonyl)amino)-4-iodobutanoate

Precaución: for Research Use Only

Formula: C₁₃H₂₄INO₄
MDL Number: MFCD27937854
CAS Number: 161370-66-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR