N-((1S,2S)-2-Amino-1,2-diphenylethyl)-2,3,4,5,6-pentafluorobenzenesulfonamide 97%

Supplier: Ambeed
Warning

A386459-1G A386459-250MG
77251-972EA 67 USD
77251-972 77251-974
N-((1S,2S)-2-Amino-1,2-diphenylethyl)-2,3,4,5,6-pentafluorobenzenesulfonamide 97%
N-((1S,2S)-2-Amino-1,2-diphenylethyl)-2,3,4,5,6-pentafluorobenzenesulfonamide

Precaución: for Research Use Only

Formula: C₂₀H₁₅F₅N₂O₂S
CAS Number: 313342-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR