4-(1-Naphthyl)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: 4-(1-Naphthyl)benzeneboronic acid

A386343-10G A386343-1G A386343-5G
77291-028EA 17 USD
77291-028 77291-030 77291-032
4-(1-Naphthyl)phenylboronic acid 97%
4-(1-Naphthyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₆H₁₃BO₂
MDL Number: MFCD08669639
CAS Number: 870774-25-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR