2-(Isopropylamino)ethanol 99%

Supplier: Ambeed
Danger

Synonyms: N-Isopropylethanolamine

A386338-5G A386338-25G
77251-970EA 16.48 USD
77251-970 77568-784
2-(Isopropylamino)ethanol 99%
2-(Isopropylamino)ethanol

Precaución: For research use only.

Formula: C₅H₁₃NO
MW: 103.17 g/mol
MDL Number: MFCD00041755
CAS Number: 109-56-8
UN: 3082
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR