Methyl-4-pentenoate 95%

Supplier: Ambeed
Danger

Synonyms: Methyl pent-4-enoate

A386002-1G A386002-5G A386002-25G
77264-308EA 20.6 USD
77264-308 77594-174 77616-676
Methyl-4-pentenoate 95%
Methyl-4-pentenoate

Precaución: for Research Use Only

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
MDL Number: MFCD03990590
CAS Number: 818-57-5
UN: 3272
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR