Kevetrin hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 3-cyanopropyl carbamimidothioate hydrochloride, Kevetrin HCl

A385793-10MG A385793-1MG A385793-25MG A385793-50MG A385793-5MG
77240-450EA 38.11 USD
77240-450 77240-452 77240-454 77240-456 77240-458
Kevetrin hydrochloride 98%
Kevetrin hydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₀ClN₃S
MW: 179.67 g/mol
MDL Number: MFCD22572767
CAS Number: 66592-89-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR