3-Chloro-5-(trifluoromethyl)pyridine 97%

Supplier: Ambeed
Warning

A385741-100G A385741-10G A385741-25G A385741-5G
77278-576EA 103 USD
77278-576 77278-578 77278-580 77278-582
3-Chloro-5-(trifluoromethyl)pyridine 97%
3-Chloro-5-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C6H3ClF3N
MW: 181.55 g/mol
MDL Number: MFCD00042227
CAS Number: 85148-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR