2-Chloro-6-(trifluoromethyl)nicotinonitrile 98%

Supplier: Ambeed
Warning

A385717-100G A385717-1G A385717-25G A385717-5G
77264-294EA 934.99 USD
77264-294 77264-296 77264-298 77264-300
2-Chloro-6-(trifluoromethyl)nicotinonitrile 98%
2-Chloro-6-(trifluoromethyl)nicotinonitrile

Precaución: for Research Use Only

Formula: C₇H₂ClF₃N₂
MW: 206.5500 g/mol
MDL Number: MFCD01862660
CAS Number: 386704-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR