trans-4-Chloro-3-nitrocinnamic acid 98%

Supplier: Ambeed
Warning

Synonyms: trans-3-(4-Chloro-3-nitrophenyl)prop-2-enoic acid, (E)-3-(4-Chloro-3-nitrophenyl)acrylic acid, trans-3-(4-Chloro-3-nitrophenyl)acrylic acid

A385662-100G A385662-1G A385662-25G A385662-5G
77264-286EA 331.66 USD
77264-286 77264-288 77264-290 77264-292
trans-4-Chloro-3-nitrocinnamic acid 98%
trans-4-Chloro-3-nitrocinnamic acid

Precaución: for Research Use Only

Formula: C₉H₆ClNO₄
MW: 227.601 g/mol
MDL Number: MFCD00063311
CAS Number: 199679-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR