3',6'-Dimethoxyfluoran 98%

Supplier: Ambeed
Warning

A385040-25G A385040-5G A385040-100G
77240-446EA 72 USD
77240-446 77240-448 77746-775
3',6'-Dimethoxyfluoran 98%
3',6'-Dimethoxyfluoran

Precaución: for Research Use Only

Formula: C₂₂H₁₆O₅
MDL Number: MFCD08276350
CAS Number: 36886-76-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR