Angelicin 98%

Supplier: Ambeed
Warning

A384656-10MG A384656-1MG A384656-5MG A384656-1G A384656-250MG
77278-570EA 20 USD
77278-570 77278-572 77278-574 77616-672 77616-674
Angelicin 98%
Angelicin

Precaución: for Research Use Only

Formula: C₁₁H₆O₃
MDL Number: MFCD00064930
CAS Number: 523-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR