4'-Chloro-2'-fluoroacetophenone 97%

Supplier: Ambeed
Warning

A384590-100G A384590-10G A384590-1G A384590-25G A384590-5G
77291-016EA 221.45 USD
77291-016 77291-018 77291-020 77291-022 77291-024
4'-Chloro-2'-fluoroacetophenone 97%
4'-Chloro-2'-fluoroacetophenone

Precaución: for Research Use Only

Formula: C₈H₆ClFO
MW: 172.59 g/mol
MDL Number: MFCD04116328
CAS Number: 175711-83-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR