2-Nitropyridine-3-carboxylic acid 97%

Supplier: Ambeed
Warning

A384497-250MG A384497-1G
77278-568EA 21 USD
77278-568 77568-772
2-Nitropyridine-3-carboxylic acid 97%
2-Nitropyridine-3-carboxylic acid

Precaución: For research use only.

Formula: C₆H₄N₂O₄
MDL Number: MFCD04114130
CAS Number: 33225-72-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR