2,2'-Oxybis(nitrobenzene) 95%

Supplier: Ambeed
Danger

A384376-100MG A384376-1G A384376-250MG A384376-25G A384376-5G
77264-276EA 23 USD
77264-276 77264-278 77264-280 77264-282 77264-284
2,2'-Oxybis(nitrobenzene) 95%
2,2'-Oxybis(nitrobenzene)

Precaución: for Research Use Only

Formula: C₁₂H₈N₂O₅
MDL Number: MFCD00033880
CAS Number: 2217-65-4
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR