Bis[2-(4,6-difluorophenyl)pyridinato-C2,N](picolinato)iridium(III) 98%

Supplier: Ambeed
Warning

A384052-100MG A384052-1G A384052-50MG A384052-250MG
77240-432EA 51 USD
77240-432 77240-434 77240-436 77746-773
Bis[2-(4,6-difluorophenyl)pyridinato-C2,N](picolinato)iridium(III) 98%
Bis[2-(4,6-difluorophenyl)pyridinato-C2,N](picolinato)iridium(III)

Precaución: for Research Use Only

Formula: C₂₈H₁₆F₄IrN₃O₂
MDL Number: MFCD09038721
CAS Number: 376367-93-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR