4-Chloro-2-methyl-7H-pyrrolo[2,3-d]pyrimidine 98%

Supplier: Ambeed
Warning

A383893-10G A383893-1G A383893-25G A383893-5G A383893-100G
77240-422EA 41 USD
77240-422 77240-424 77240-426 77240-428 77240-430 77568-770
4-Chloro-2-methyl-7H-pyrrolo[2,3-d]pyrimidine 98%
4-Chloro-2-methyl-7H-pyrrolo[2,3-d]pyrimidine

Precaución: For research use only.

Formula: C₇H₆ClN₃
MDL Number: MFCD09746275
CAS Number: 71149-52-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR