Bis(4-pyridyl)amine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
4,4'-Dipyridylamine, N-(4-Pyridinyl)-4-pyridinamine, Di(pyridin-4-yl)amine
A383818-100MG
A383818-1G
A383818-250MG
A383818-25G
A383818-5G
77290-994EA
6.18
USD
77290-994
77290-996
77290-998
77291-000
77291-002
Bis(4-pyridyl)amine 95%
Bis(4-pyridyl)amine
Precaución: for Research Use Only
Formula:
C₁₀H₉N₃ MW: 171.20 g/mol |
MDL Number:
MFCD00829434 CAS Number: 1915-42-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...