3-Chloro-2-(difluoromethyl)pyridine 97%

Supplier: Ambeed
Warning

A383791-100MG A383791-250MG
77264-270EA 100.94 USD
77264-270 77264-272
3-Chloro-2-(difluoromethyl)pyridine 97%
3-Chloro-2-(difluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₄ClF₂N
MW: 163.55 g/mol
MDL Number: MFCD25476534
CAS Number: 1805301-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR