4-(2-Methoxy-2-oxoethoxy)benzeneboronic acid, pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-[4-(tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]acetate, Methyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy)acetate

A383718-1G A383718-250MG A383718-5G
77240-416EA 87 USD
77240-416 77240-418 77240-420
4-(2-Methoxy-2-oxoethoxy)benzeneboronic acid, pinacol ester 97%
4-(2-Methoxy-2-oxoethoxy)benzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₁BO₅
MW: 292.135 g/mol
MDL Number: MFCD27957319
CAS Number: 1610517-55-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR