3-Chloro-4-(trifluoromethoxy)benzyl alcohol 98%

Supplier: Ambeed
Warning

Synonyms: [3-Chloro-4-(trifluoromethoxy)phenyl]methanol, (3-Chloro-4-(trifluoromethoxy)phenyl)methanol

A383390-1G A383390-250MG
77278-564EA 37.08 USD
77278-564 77278-566
3-Chloro-4-(trifluoromethoxy)benzyl alcohol 98%
3-Chloro-4-(trifluoromethoxy)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆ClF₃O₂
MW: 226.58 g/mol
MDL Number: MFCD01631559
CAS Number: 56456-48-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR