3-Bromo-2,6-difluorobenzaldehyde 98%

Supplier: Ambeed
Warning

A383383-1G A383383-250MG A383383-25G A383383-5G
77278-556EA 10 USD
77278-556 77278-558 77278-560 77278-562
3-Bromo-2,6-difluorobenzaldehyde 98%
3-Bromo-2,6-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O
MDL Number: MFCD08282773
CAS Number: 398456-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR