t-Boc-N-Amido-PEG6-Amine 95%

Supplier: Ambeed
Warning

Synonyms: Boc-NH-PEG6-amine, Boc-NH-PEG6-NH2

A383378-100MG A383378-250MG A383378-1G A383378-50MG
77240-412EA 59 USD
77240-412 77240-414 77551-120 77746-770
t-Boc-N-Amido-PEG6-Amine 95%
t-Boc-N-Amido-PEG6-Amine

Precaución: for Research Use Only

Formula: C₁₉H₄₀N₂O₈
MDL Number: MFCD16619391
CAS Number: 1091627-77-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR