2-(1H-Imidazol-2-yl)propan-2-ol 97%

Supplier: Ambeed
Warning

A383187-250MG A383187-1G
77240-410EA 131.84 USD
77240-410 77746-769
2-(1H-Imidazol-2-yl)propan-2-ol 97%
2-(1H-Imidazol-2-yl)propan-2-ol

Precaución: for Research Use Only

Formula: C₆H₁₀N₂O
MW: 126.15 g/mol
MDL Number: MFCD19218235
CAS Number: 36365-23-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR