2-Chloro-5-phenylthiazole 95%

Supplier: Ambeed
Warning

A382734-100MG A382734-1G
77264-254EA 76.22 USD
77264-254 77264-256
2-Chloro-5-phenylthiazole 95%
2-Chloro-5-phenylthiazole

Precaución: for Research Use Only

Formula: C₉H₆ClNS
MW: 195.66 g/mol
MDL Number: MFCD08236828
CAS Number: 329794-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR