4-Acetyl-3-fluorophenylboronic acid 98%

Supplier: Ambeed
Warning

A382422-1G A382422-250MG A382422-5G
77278-546EA 36.05 USD
77278-546 77278-548 77278-550
4-Acetyl-3-fluorophenylboronic acid 98%
4-Acetyl-3-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BFO₃
MW: 181.9600 g/mol
MDL Number: MFCD06796422
CAS Number: 481725-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR