4-Cyano-4'-hexyloxybiphenyl 98%

Supplier: Ambeed
Warning

A382030-100G A382030-1G A382030-25G A382030-5G
77290-986EA 275 USD
77290-986 77290-988 77290-990 77290-992
4-Cyano-4'-hexyloxybiphenyl 98%
4-Cyano-4'-hexyloxybiphenyl

Precaución: for Research Use Only

Formula: C₁₉H₂₁NO
MDL Number: MFCD00134145
CAS Number: 41424-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR