(R)-2,2,2-Trifluoro-1-(4-(trifluoromethyl)phenyl)ethanamine hydrochloride 96%

Supplier: Ambeed
Warning

A381781-100MG A381781-250MG
77264-250EA 70.04 USD
77264-250 77264-252
(R)-2,2,2-Trifluoro-1-(4-(trifluoromethyl)phenyl)ethanamine hydrochloride 96%
(R)-2,2,2-Trifluoro-1-(4-(trifluoromethyl)phenyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₈ClF₆N
MW: 279.60 g/mol
MDL Number: MFCD12757352
CAS Number: 1260618-04-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR