tert-Butyl 4-(4-bromo-2-nitrophenyl)piperazine-1-carboxylate 97%

Supplier: Ambeed
Warning

A381595-100MG A381595-1G A381595-250MG A381595-5G
77240-400EA 37.08 USD
77240-400 77240-402 77240-404 77240-406
tert-Butyl 4-(4-bromo-2-nitrophenyl)piperazine-1-carboxylate 97%
tert-Butyl 4-(4-bromo-2-nitrophenyl)piperazine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₀BrN₃O₄
MW: 386.24 g/mol
MDL Number: MFCD11041977
CAS Number: 474329-57-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR