2-Chloro-1,1,1-trimethoxyethane 98% (by GC)

Supplier: Ambeed
Danger

A381525-25G A381525-5G
77264-248EA 33.99 USD
77264-248 77572-132
2-Chloro-1,1,1-trimethoxyethane 98% (by GC)
2-Chloro-1,1,1-trimethoxyethane

Precaución: for Research Use Only

Formula: C₅H₁₁ClO₃
MW: 154.59 g/mol
MDL Number: MFCD00216629
CAS Number: 74974-54-2
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR