(E)-2-(3-(Naphthalen-2-yl)but-2-enamido)benzoic acid 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
BIBR 1532
A381410-100MG
A381410-10MG
A381410-1MG
A381410-50MG
A381410-5MG
A381410-1G
A381410-250MG
77240-390EA
138.02
USD
77240-390
77240-392
77240-394
77240-396
77240-398
77746-766
77746-767
(E)-2-(3-(Naphthalen-2-yl)but-2-enamido)benzoic acid 99%
(E)-2-(3-(Naphthalen-2-yl)but-2-enamido)benzoic acid
Precaución: for Research Use Only
Formula:
C₂₁H₁₇NO₃ MW: 331.36 g/mol |
MDL Number:
MFCD11112195 CAS Number: 321674-73-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...