(R)-3,3'-Di-9-phenanthrenyl-1,1'-binaphthalene-2,2'-diol ≥98%, ee 99%
Supplier: Ambeed
|
Warning
|
A380940-100MG
A380940-1G
A380940-250MG
A380940-5G
77251-880EA
135.96
USD
77251-880
77251-882
77251-884
77251-886
(R)-3,3'-Di-9-phenanthrenyl-1,1'-binaphthalene-2,2'-diol ≥98%, ee 99%
(R)-3,3'-Di-9-phenanthrenyl-1,1'-binaphthalene-2,2'-diol
Precaución: for Research Use Only
Formula:
C₄₈H₃₀O₂ MW: 638.75 g/mol |
MDL Number:
MFCD09027269 CAS Number: 1058734-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...