4-(2-Methylimidazol-1-yl)phenylamine 95%

Supplier: Ambeed
Warning

Synonyms: 4-(2-Methyl-1H-imidazol-1-yl)aniline

A380902-100MG A380902-25G A380902-5G
77290-980EA 11.33 USD
77290-980 77290-982 77290-984
4-(2-Methylimidazol-1-yl)phenylamine 95%
4-(2-Methylimidazol-1-yl)phenylamine

Precaución: for Research Use Only

Formula: C₁₀H₁₁N₃
MW: 173.2144 g/mol
MDL Number: MFCD06797788
CAS Number: 74852-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR