4-Bromo-1-methyl-1H-pyrazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A380640-250MG A380640-1G
77290-976EA 9.27 USD
77290-976 77748-762
4-Bromo-1-methyl-1H-pyrazole-3-carboxylic acid 95%
4-Bromo-1-methyl-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O₂
MDL Number: MFCD00463988
CAS Number: 84547-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR