1-Phenylcyclobutanecarbonitrile 95%

Supplier: Ambeed
Warning

A380418-100G A380418-10G A380418-1G A380418-25G A380418-5G
77240-372EA 378 USD
77240-372 77240-374 77240-376 77240-378 77240-380
1-Phenylcyclobutanecarbonitrile 95%
1-Phenylcyclobutanecarbonitrile

Precaución: for Research Use Only

Formula: C₁₁H₁₁N
MDL Number: MFCD00019258
CAS Number: 14377-68-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR