Adenosine, N-benzoyl-5'-O-[bis(4-methoxyphenyl)phenylmethyl]-3'-O-[(1,1-dimethylethyl)dimethylsilyl]-, 2'-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite] 97%

Supplier: Ambeed
Warning

Synonyms: 3'-TBDMS-Bz-rA Phosphoramidite

A380332-1G A380332-5G
77264-236EA 80 USD
77264-236 77264-238
Adenosine, N-benzoyl-5'-O-[bis(4-methoxyphenyl)phenylmethyl]-3'-O-[(1,1-dimethylethyl)dimethylsilyl]-, 2'-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite] 97%
Adenosine, N-benzoyl-5'-O-[bis(4-methoxyphenyl)phenylmethyl]-3'-O-[(1,1-dimethylethyl)dimethylsilyl]-, 2'-[2-cyanoethyl N,N-bis(1-methylethyl)phosphoramidite]

Precaución: for Research Use Only

Formula: C₅₃H₆₆N₇O₈PSi
MW: 988.19 g/mol
MDL Number: MFCD00274109
CAS Number: 129451-75-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR