Methyl 7-fluoro-1H-indole-6-carboxylate 97%

Supplier: Ambeed
Warning

A380278-100MG A380278-1G A380278-250MG
77264-228EA 128.75 USD
77264-228 77264-230 77264-232
Methyl 7-fluoro-1H-indole-6-carboxylate 97%
Methyl 7-fluoro-1H-indole-6-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₈FNO₂
MW: 193.17 g/mol
MDL Number: MFCD23706323
CAS Number: 1427366-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR