α,3-Dibromo-5-fluorotoluene 95%

Supplier: Ambeed
Danger

Synonyms: 3-Bromo-5-fluorobenzyl bromide, 1-Bromo-3-(bromomethyl)-5-fluorobenzene

A379913-250MG A379913-5G
77278-522EA 5.15 USD
77278-522 77278-524
α,3-Dibromo-5-fluorotoluene 95%
α,3-Dibromo-5-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅Br₂F
MW: 267.92 g/mol
MDL Number: MFCD09258959
CAS Number: 216755-57-6
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR