Methyl 3-amino-5,6-dihydro-4H-thieno[2,3-c]pyrrole-2-carboxylate dihydrochloride 97%

Supplier: Ambeed
Warning

A379752-100MG A379752-250MG
77240-362EA 91.67 USD
77240-362 77240-364
Methyl 3-amino-5,6-dihydro-4H-thieno[2,3-c]pyrrole-2-carboxylate dihydrochloride 97%
Methyl 3-amino-5,6-dihydro-4H-thieno[2,3-c]pyrrole-2-carboxylate dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂Cl₂N₂O₂S
MW: 271.16 g/mol
MDL Number: MFCD30478568
CAS Number: 1998216-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR