(S)-3,3'-Bis(2-naphthalenyl)-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate 98%

Supplier: Ambeed
Warning

A379467-100MG A379467-1G A379467-250MG
77251-850EA 57 USD
77251-850 77251-852 77251-854
(S)-3,3'-Bis(2-naphthalenyl)-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate 98%
(S)-3,3'-Bis(2-naphthalenyl)-1,1'-binaphthyl-2,2'-diyl Hydrogen Phosphate

Precaución: for Research Use Only

Formula: C₄₀H₂₅O₄P
CAS Number: 874948-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR