3-Chloro-2-iodophenol 97%

Supplier: Ambeed
Warning

A379289-100MG A379289-250MG A379289-5G
77278-512EA 17 USD
77278-512 77278-514 77748-759
3-Chloro-2-iodophenol 97%
3-Chloro-2-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄ClIO
MDL Number: MFCD08166414
CAS Number: 858854-82-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR