3-(sulfanylmethyl)pyridine 95%

Supplier: Ambeed
Danger

Synonyms: 3-Pyridinemethanethiol, 3-(Mercaptomethyl)pyridine, (Pyridin-3-yl)methanethiol

A379276-1G A379276-250MG A379276-5G
77290-966EA 468.65 USD
77290-966 77290-968 77290-970
3-(sulfanylmethyl)pyridine 95%
3-(sulfanylmethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₇NS
MW: 125.191 g/mol
MDL Number: MFCD01691719
CAS Number: 17617-05-9
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR