2-(Boc-amino)-3-methylpyridine 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(N-Boc-Amino)-3-methylpyridine
A379215-100G
A379215-10G
A379215-1G
A379215-25G
A379215-5G
77240-352EA
255.44
USD
77240-352
77240-354
77240-356
77240-358
77240-360
2-(Boc-amino)-3-methylpyridine 97%
2-(Boc-amino)-3-methylpyridine
Precaución: for Research Use Only
Formula:
C₁₁H₁₆N₂O₂ MW: 208.26 g/mol |
MDL Number:
MFCD03425358 CAS Number: 138343-75-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...