2-Methoxyphenethyl bromide 97%

Supplier: Ambeed
Warning

Synonyms: 1-(2-Bromoethyl)-2-methoxybenzene

A379185-1G A379185-5G
77264-224EA 104.03 USD
77264-224 77264-226
2-Methoxyphenethyl bromide 97%
2-Methoxyphenethyl bromide

Precaución: for Research Use Only

Formula: C₉H₁₁BrO
MW: 215.09 g/mol
MDL Number: MFCD07369785
CAS Number: 36449-75-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR