2,4,5-Tribromoimidazole 98%

Supplier: Ambeed
Danger

A379080-100G A379080-10G
77290-962EA 103 USD
77290-962 77290-964
2,4,5-Tribromoimidazole 98%
2,4,5-Tribromoimidazole

Precaución: for Research Use Only

Formula: C₃HBr₃N₂
MDL Number: MFCD00005184
CAS Number: 2034-22-2
UN: 2811
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR