(S)-Ethyl-2-((S)-4-methyl-2,5-dioxooxazolidin-3-yl)-4-phenylbutanoate 97%

Supplier: Ambeed
Warning

Synonyms: (S)-2-[(S)-4-Methyl-2,5-dioxooxazolidin-3-yl]-4-phenylbutanoic acid ethyl ester, Ethyl (S)-2-[(S)-4-Methyl-2,5-dioxooxazolidin-3-yl]-4-phenylbutanoate, Ethyl (S)-2-[(S)-4-Methyl-2,5-dioxo-3-oxazolidinyl]-4-phenylbutanoate, N-[1-(S)-Ethoxycarbonyl-3-phenylpropyl]-L-alanine-N-carboxyanhydride

A378827-100G A378827-10G
77278-508EA 75.19 USD
77278-508 77278-510
(S)-Ethyl-2-((S)-4-methyl-2,5-dioxooxazolidin-3-yl)-4-phenylbutanoate 97%
(S)-Ethyl-2-((S)-4-methyl-2,5-dioxooxazolidin-3-yl)-4-phenylbutanoate

Precaución: for Research Use Only

Formula: C₁₆H₁₉NO₅
MW: 305.33 g/mol
MDL Number: MFCD00272296
CAS Number: 84793-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR