4-(4-Fluorophenyl)butan-2-one 97%

Supplier: Ambeed
Warning

A378415-100MG A378415-1G A378415-250MG
77264-210EA 95.79 USD
77264-210 77264-212 77264-214
4-(4-Fluorophenyl)butan-2-one 97%
4-(4-Fluorophenyl)butan-2-one

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO
MW: 166.19 g/mol
MDL Number: MFCD09891020
CAS Number: 63416-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR