3-Bromo-2,4,5-trifluoroaniline 95%

Supplier: Ambeed
Warning

A378103-1G A378103-25G A378103-5G
77264-204EA 26.78 USD
77264-204 77264-206 77264-208
3-Bromo-2,4,5-trifluoroaniline 95%
3-Bromo-2,4,5-trifluoroaniline

Precaución: for Research Use Only

Formula: C₆H₃BrF₃N
MW: 225.994 g/mol
MDL Number: MFCD30530500
CAS Number: 2055841-42-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR