4'-(Trifluoromethyl)acetanilide 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-(Trifluoromethyl)phenyl)acetamide, 4'-(Trifluoromethyl)acetanilide, 4-Acetamidobenzotrifluoride, alpha,alpha,alpha-Trifluoro-p-acetotoluidide

A377619-100G A377619-25G A377619-5G
77290-946EA 224.54 USD
77290-946 77290-948 77290-950
4'-(Trifluoromethyl)acetanilide 98%
4'-(Trifluoromethyl)acetanilide

Precaución: for Research Use Only

Formula: C₉H₈F₃NO
MW: 203.16 g/mol
MDL Number: MFCD00013562
CAS Number: 349-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR