6-Methylpyrazin-2-ol 98%

Supplier: Ambeed
Warning

Synonyms: 2-Hydroxy-6-methylpyrazine

A377229-100G A377229-1G
77264-182EA 394.49 USD
77264-182 77264-184
6-Methylpyrazin-2-ol 98%
6-Methylpyrazin-2-ol

Precaución: for Research Use Only

Formula: C₅H₆N₂O
MW: 110.1139 g/mol
MDL Number: MFCD08705761
CAS Number: 20721-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR