4',5'-Methylenedioxy-2'-nitroacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(6-Nitrobenzo[d][1,3]dioxol-5-yl)ethanone

A377037-1G A377037-250MG A377037-5G
77264-178EA 30.9 USD
77264-178 77264-180 77594-132
4',5'-Methylenedioxy-2'-nitroacetophenone 98%
4',5'-Methylenedioxy-2'-nitroacetophenone

Precaución: for Research Use Only

Formula: C₉H₇NO₅
MW: 209.16 g/mol
MDL Number: MFCD00053045
CAS Number: 56136-84-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR